Learn More
Biomatik Corporation Human Tyrosinase (TYR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35949
Encompass_Preferred
Human Tyrosinase (Tyr) Elisa Kit for detecting Tyrosinase (Oculocutaneous Albinism Ia) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P14679; TYR. Target Synonyms: ATN; CMM8; LB24 AB; LB24-AB; Monophenol monooxygenase; OCA1; OCA1A; OCAIA; Oculocutaneous albinism IA; SHEP3; SK29 AB; SK29-AB; Tumor rejection antigen AB; TYR; TYRO_HUMAN; tyrosinase (oculocutaneous albinism IA); Tyrosinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8343