Learn More
Biomatik Corporation Human Tyrosine-protein kinase ABL1 (ABL1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35953
Encompass_Preferred
Human Tyrosine-Protein Kinase Abl1 (Abl1) Elisa Kit for detecting C-Abl Oncogene 1, Receptor Tyrosine Kinase in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 25-1600pg/ml. Sensitivity: 6.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P00519; ABL1. Target Synonyms: Abelson murine leukemia viral oncogene homolog 1; Abelson tyrosine protein kinase 1; Abl 1; ABL; ABL proto oncogene 1 non receptor tyrosine kinase; ABL1; ABL1_HUMAN; bcr/abl; bcr/c abl oncogene protein; c ABL; c abl oncogene 1 non receptor tyrosine kinase; c abl oncogene 1 receptor tyrosine kinase; c ABL1; JTK7; p150; Proto oncogene tyrosine protein kinase ABL1; Proto-oncogene c-Abl; Tyrosine-protein kinase ABL1; v abl Abelson murine leukemia viral oncogene homolog 1; v abl. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8344