Learn More
Biomatik Corporation Human Uncharacterized protein C9orf72 (C9orf72) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC35992
Encompass_Preferred
Human Uncharacterized Protein C9Orf72 (C9Orf72) Elisa Kit for detecting Chromosome 9 Open Reading Frame 72 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q96LT7; C9orf72. Target Synonyms: ALSFTD; C9orf72; chromosome 9 open reading frame 72; CI072_HUMAN; FTDALS; MGC23980; Protein C9orf72; RP11-27J8.2; Uncharacterized protein C9orf72. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8349