Learn More
Biomatik Corporation Human upper zone of growth plate and cartilage matrix associated (UCMA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36000
Encompass_Preferred
Human Upper Zone Of Growth Plate And Cartilage Matrix Associated (Ucma) Elisa Kit for detecting Upper Zone Of Growth Plate And Cartilage Matrix Associated in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8WVF2; UCMA. Target Synonyms: UCMA; C10orf49; Unique cartilage matrix-associated protein [Cleaved into: Unique cartilage matrix-associated protein C-terminal fragment; Ucma-C; Gla-rich protein; GRP)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8351