Learn More
Biomatik Corporation Human Vitamin D 25-hydroxylase (CYP2R1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36034
Encompass_Preferred
Human Vitamin D 25-Hydroxylase (Cyp2R1) Elisa Kit for detecting Cytochrome P450, Family 2, Subfamily R, Polypeptide 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q6VVX0; CYP2R1. Target Synonyms: CP2R1_HUMAN; CYP2R1; Cytochrome P450 2R1; Cytochrome P450 family 2 R1; Cytochrome P450 family 2 subfamily R polypeptide 1; EC 1.14.13.15; MGC4663; Vitamin D 25 hydroxylase; Vitamin D 25-hydroxylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8355