Learn More
Biomatik Corporation Human Vitamin D-binding protein, DBP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36036
Encompass_Preferred
Human Vitamin D-Binding Protein, Dbp Elisa Kit for detecting Vitamin D-Binding Protein, Dbp in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.3125-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P02774; GC. Target Synonyms: DBP; DBP/GC; GC; Gc globulin; Gc-globulin; GRD3; Group specific component; Group specific component vitamin D binding protein; Group-specific component; hDBP; VDB; VDBG; VDBP; Vitamin D binding alpha globulin; Vitamin D-binding protein; VTDB_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8357