Learn More
Biomatik Corporation Human V-set and immunoglobulin domain-containing protein 4 (VSIG4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36049
Encompass_Preferred
Human V-Set And Immunoglobulin Domain-Containing Protein 4 (Vsig4) Elisa Kit for detecting V-Set And Immunoglobulin Domain Containing 4 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9Y279; VSIG4. Target Synonyms: Complement receptor of the immunoglobulin superfamily; CRIg; Ig superfamily protein; Protein Z39Ig; UNQ317/PRO362; V set and immunoglobulin domain containing 4; V-set and immunoglobulin domain-containing protein 4; VSIG4; VSIG4_HUMAN; Z39IG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8358