Learn More
Biomatik Corporation Human zonulin ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36091
Encompass_Preferred
Human Zonulin Elisa Kit for detecting Zonulin in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8363