Learn More
Biomatik Corporation Human Zyxin (ZYX) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36093
Encompass_Preferred
Human Zyxin (Zyx) Elisa Kit for detecting Zyxin in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q15942; ZYX. Target Synonyms: ZYX; Zyxin; Zyxin-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8364