Learn More
Biomatik Corporation Human α1-Antitrypsin, α1-AT ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36095
Encompass_Preferred
The Human α1-Antitrypsin, α1-AT ELISA Kit allows for quantitative determination of serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1 in serum, plasma. Species Reactivity: Human. Detection Range: 78 nIU/mL-5000 nIU/mL. Sensitivity: 19.5 nIU/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P01009. Restriction: For Research Use Only.
Catalog No. 50-149-8365