Learn More
Biomatik Corporation Human β2-microglobulin, BMG/β2-MG ELISA Kit, 96 tests
Catalog No. 501498369
Encompass_Preferred
The Human β2-microglobulin, BMG/β2-MG ELISA Kit allows for quantitative determination of beta-2-microglobulin in serum, urine. Species Reactivity: Human. Detection Range: 0.025 μg/mL-10 μg/mL. Sensitivity: 0.01 μg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P61769. Restriction: For Research Use Only.
Catalog No. 50-149-8369 Supplier Biomatik Corporation Supplier No. EKC36111
The Human β2-microglobulin, BMG/β2-MG ELISA Kit allows for quantitative determination of beta-2-microglobulin in serum, urine. Species Reactivity: Human. Detection Range: 0.025 μg/mL-10 μg/mL. Sensitivity: 0.01 μg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P61769. Restriction: For Research Use Only.