Learn More
Biomatik Corporation Human β-defensins 1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36114
Encompass_Preferred
The Human β-defensins 1 ELISA Kit allows for quantitative determination of defensin, beta 1 in serum, plasma, tissue homogenates. Species Reactivity: Human. Detection Range: 3.12 pg/mL-200 pg/mL. Sensitivity: 0.78 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P60022. Restriction: For Research Use Only.
Catalog No. 50-149-8371