Learn More
Biomatik Corporation Human β-subunits of hemoglobin, HB-β ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36122
Encompass_Preferred
The Human β-subunits of hemoglobin, HB-β ELISA Kit allows for quantitative determination of hemoglobin, beta in serum, plasma, lysate for RBC. Species Reactivity: Human. Detection Range: 0.12 ng/mL-500 ng/mL. Sensitivity: 0.12 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P68871. Restriction: For Research Use Only.
Catalog No. 50-149-8372