Learn More
Biomatik Corporation Human γ-Secretase ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36125
Encompass_Preferred
The Human γ-Secretase ELISA Kit allows for quantitative determination of γ-Secretase in serum, plasma and tissue homogenates. Species Reactivity: Human. Detection Range: 31.25 pg/mL-2000 pg/mL. Sensitivity: 7.81 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-8373