Learn More
Biomatik Corporation Monkey Apolipoprotein (a) (LPA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36135
Encompass_Preferred
Monkey Apolipoprotein (A) (Lpa) Elisa Kit for detecting Lipoprotein, Lp(A) in serum, plasma, tissue homogenates. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 31.25-2000ng/ml. Sensitivity: 15.63ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P14417; LPA. Target Synonyms: LPAApolipoprotein (a; Apo (a); Lp (a); EC 3.4.21.-; Fragment. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8375