Learn More
Biomatik Corporation Monkey complement C5a anaphylatoxin (C5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36148
Encompass_Preferred
Monkey Complement C5 (C5) Elisa Kit for detecting Complement C5A Anaphylatoxin in serum, plasma, tissue homogenates. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: CPAMD4; FLJ17816; FLJ17822; MGC142298; anaphylatoxin C5a analog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8377