Learn More
Biomatik Corporation Monkey cortisol (COR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36149
Encompass_Preferred
Monkey Cortisol (Cor) Elisa Kit for detecting Cortisol(Cor) in serum, plasma, tissue homogenates. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 0.049-200ng/ml. Sensitivity: 0.049ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8378