Learn More
Biomatik Corporation Monkey C-X-C motif chemokine 10 (CXCL10) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36150
Encompass_Preferred
Monkey C-X-C Motif Chemokine 10 (Cxcl10) Elisa Kit for detecting Chemokine (C-X-C Motif) Ligand 10 in serum, plasma, tissue homogenates. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8MIZ1; CXCL10. Target Synonyms: CXCL10; SCYB10C-X-C motif chemokine 10; 10 kDa interferon gamma-induced protein; Gamma-IP10; IP-10; Small-inducible cytokine B10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8379