Learn More
Biomatik Corporation Monkey hepatitis A virus cellular receptor 1/KIM1 (HAVCR1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36163
Encompass_Preferred
Monkey Hepatitis A Virus Cellular Receptor 1/Kim1 (Havcr1) Elisa Kit for detecting Hepatitis A Virus Cellular Receptor 1/Kim1(Havcr1) in serum, urine. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O46598; HAVCR1. Target Synonyms: HAVCR1; TIM1; TIMD1; Hepatitis A virus cellular receptor 1; HAVcr-1; T-cell immunoglobulin and mucin domain-containing protein 1; TIMD-1; T-cell immunoglobulin mucin receptor 1; TIM-1; T-cell membrane protein 1; CD antigen CD365. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8384