Learn More
Biomatik Corporation Monkey Insulin-like Growth Factor 1 (IGF-1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36170
Encompass_Preferred
Monkey Insulin-Like Growth Factor 1 (Igf-1) Elisa Kit for detecting Insulin-Like Growth Factor 1(Igf-1) in serum, plasma. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 62.5-4000ng/ml. Sensitivity: 15.6ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q68LC0; IGF1. Target Synonyms: IGF1; Insulin-like growth factor I; IGF-I; Somatomedin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8385