Learn More
Biomatik Corporation Monkey Interleukin-1 alpha (IL1A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36178
Encompass_Preferred
Monkey Interleukin-1 Alpha (Il1A) Elisa Kit for detecting Interleukin 1, Alpha in serum, plasma, tissue homogenates. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P48089; IL1A. Target Synonyms: IL1A; Interleukin-1 alpha; IL-1 alpha; Hematopoietin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8386