Learn More
Biomatik Corporation Monkey Leptin, LEP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36182
Encompass_Preferred
Monkey Leptin, Lep Elisa Kit for detecting Leptin in serum, plasma, tissue homogenates. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 3.125-50ng/ml. Sensitivity: 3.125ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q28504; LEP. Target Synonyms: LEP; OB; Leptin; Obesity factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8387