Learn More
Biomatik Corporation Monkey Proprotein convertase subtilisin/kexin type 9 (PCSK9) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36195
Encompass_Preferred
Monkey Proprotein Convertase Subtilisin/Kexin Type 9 (Pcsk9) Elisa Kit for detecting Proprotein Convertase Subtilisin/Kexin Type 9 in serum, plasma, tissue homogenates. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 1.1-800pg/ml. Sensitivity: 1.1pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: A8T666; PCSK9. Target Synonyms: PCSK9; Proprotein convertase subtilisin/kexin type 9; EC 3.4.21.-; Proprotein convertase 9; PC9; Subtilisin/kexin-like protease PC9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8389