Learn More
Biomatik Corporation Monkey Tumor necrosis factor , TNF- ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36203
Encompass_Preferred
Monkey Tumor Necrosis Factor Α, Tnf-Α Elisa Kit for detecting Tumor Necrosis Factor (Tnf Superfamily, Member 2) in serum, plasma, tissue homogenates. Species Reactivity: Monkey (Simiiformes; infraorder). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P48094; TNF. Target Synonyms: TNF; TNFA; TNFSF2; Tumor necrosis factor; Cachectin; TNF-alpha; Tumor necrosis factor ligand superfamily member 2; TNF-a. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8391