Learn More
Biomatik Corporation Mouse 4-Hydroxynonenal (HNE) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36215
Encompass_Preferred
Mouse 4-Hydroxynonenal (Hne) Elisa Kit for detecting 4-Hydroxynonenal (Hne) in serum, plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pmol/ml. Sensitivity: 3.9pmol/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8395