Learn More
Biomatik Corporation Mouse Acetylcholinesterase (AChE) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36238
Encompass_Preferred
Mouse Acetylcholinesterase (Ache) Elisa Kit for detecting Acetylcholinesterase(Ache) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20mU/ml. Sensitivity: 0.078mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P21836; Ache. Target Synonyms: Ache; Acetylcholinesterase; AChE; EC 3.1.1.7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8400