Learn More
Biomatik Corporation Mouse Alanine Aminotransferase (ALT/GPT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36274
Encompass_Preferred
Mouse Alanine Aminotransferase (Alt/Gpt) Elisa Kit for detecting Alanine Aminotransferase(Alt/Gpt) in serum, plasma, cell culture supernates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10mU/ml. Sensitivity: 0.039mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8QZR5; Gpt. Target Synonyms: Gpt; Gpt1Alanine aminotransferase 1; ALT1; EC 2.6.1.2; Glutamate pyruvate transaminase 1; GPT 1; Glutamic--alanine transaminase 1; Glutamic--pyruvic transaminase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8405