Learn More
Biomatik Corporation Mouse Angiopoietin 1, ANG-1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36307
Encompass_Preferred
Mouse Angiopoietin 1, Ang-1 Elisa Kit for detecting Angiopoietin 1 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.25-80ng/ml. Sensitivity: 0.312ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O08538; Angpt1. Target Synonyms: Angpt1; AgptAngiopoietin-1; ANG-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8407