Learn More
Biomatik Corporation Mouse Angiopoietin 2, ANG-2 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36308
Encompass_Preferred
Mouse Angiopoietin 2, Ang-2 Elisa Kit for detecting Angiopoietin 2 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10pg/ml. Sensitivity: 0.039pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O35608; Angpt2. Target Synonyms: Angpt2; Agpt2Angiopoietin-2; ANG-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8408