Learn More
Biomatik Corporation Mouse Angiopoietin-related protein 4 (ANGPTL4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36312
Encompass_Preferred
Mouse Angiopoietin-Related Protein 4 (Angptl4) Elisa Kit for detecting Angiopoietin-Like 4 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q9Z1P8; Angptl4. Target Synonyms: Angptl4; Farp; Fiaf; Ng27Angiopoietin-related protein 4; 425O18-1; Angiopoietin-like protein 4; Fasting-induced adipose factor; Hepatic fibrinogen/angiopoietin-related protein; HFARP; Secreted protein Bk89) [Cleaved into: ANGPTL4 N-terminal chain; ANGPTL4 C-terminal chain]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8409