Learn More
Biomatik Corporation Mouse Angiopoietin-related protein 6 (ANGPTL6) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36313
Encompass_Preferred
Mouse Angiopoietin-Related Protein 6 (Angptl6) Elisa Kit for detecting Angiopoietin-Like 6 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8R0Z6; Angptl6. Target Synonyms: Angptl6; AgfAngiopoietin-related protein 6; Angiopoietin-like protein 6; Angiopoietin-related growth factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8410