Learn More
Biomatik Corporation Mouse Annexin A1 (ANXA1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36323
Encompass_Preferred
Mouse Annexin A1 (Anxa1) Elisa Kit for detecting Annexin A1(Anxa1) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.37-1000ng/ml. Sensitivity: 1.37ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P10107; Anxa1. Target Synonyms: Anxa1; Anx1; Lpc-1; Lpc1Annexin A1; Annexin I; Annexin-1; Calpactin II; Calpactin-2; Chromobindin-9; Lipocortin I; Phospholipase A2 inhibitory protein; p35. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8413