Learn More
Biomatik Corporation Mouse antidiuretic hormone/vasopressin/arginine vasopressin, ADH/VP/AVP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36330
Encompass_Preferred
Mouse Antidiuretic Hormone/Vasopressin/Arginine Vasopressin, Adh/Vp/Avp Elisa Kit for detecting Arginine Vasopressin in serum, plasma, cell culture supernates. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.25-80pg/ml. Sensitivity: 0.31pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: ADH; ARVP; AVP-NPII; AVRP; VP; antidiuretic hormone; arginine vasopressin-neurophysin II; neurohypophyseal; vasopressin-neurophysin II-copeptin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8414