Learn More
Biomatik Corporation Mouse Anti-Smooth Muscle Antibody, ASMA ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36343
Encompass_Preferred
Mouse Anti-Smooth Muscle Antibody, Asma Elisa Kit for detecting Anti-Smooth Muscle Antibody, Asma in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8417