Learn More
Biomatik Corporation Mouse anti-thyroid-globulin antibody, TGAB ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36345
Encompass_Preferred
Mouse Anti-Thyroid-Globulin Antibody, Tgab Elisa Kit for detecting Anti-Thyroid-Globulin Antibody, Tgab in serum, plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.25-20U/ml. Sensitivity: 1.25u/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8418