Learn More
Biomatik Corporation Mouse Apolipoprotein C-III (APOC3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36356
Encompass_Preferred
Mouse Apolipoprotein C-Iii (Apoc3) Elisa Kit for detecting Apolipoprotein C-Iii in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 18.75-1200ng/ml. Sensitivity: 9.375ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P33622; Apoc3. Target Synonyms: Apoc3Apolipoprotein C-III; Apo-CIII; ApoC-III; Apolipoprotein C3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8422