Learn More
Biomatik Corporation Mouse Arginase-1 (ARG1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36367
Encompass_Preferred
Mouse Arginase-1 (Arg1) Elisa Kit for detecting Arginase, Liver in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q61176; Arg1. Target Synonyms: Arg1Arginase-1; EC 3.5.3.1; Liver-type arginase; Type I arginase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8424