Learn More
Biomatik Corporation Mouse Atrial Natriuretic Peptide, ANP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36379
Encompass_Preferred
Mouse Atrial Natriuretic Peptide, Anp Elisa Kit for detecting Natriuretic Peptide Precursor A in serum, plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P05125; Nppa. Target Synonyms: Nppa; PndNatriuretic peptides A; Prepronatriodilatin) [Cleaved into: Atrial natriuretic factor; ANF; Atrial natriuretic peptide; ANP); Auriculin-B; Auriculin-A; Atriopeptin-1; Atriopeptin I); Atriopeptin-2; Atriopeptin II)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8427