Learn More
Biomatik Corporation mouse Beta-Endorphin, -EP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36404
Encompass_Preferred
Mouse Beta-Endorphin, Β-Ep Elisa Kit for detecting Beta-Endorphin, Β-Ep in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50pg/ml. Sensitivity: 0.195pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: beta-Endorphin; beta-EP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8430