Learn More
Biomatik Corporation Mouse Bone marrow proteoglycan (PRG2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36413
Encompass_Preferred
Mouse Bone Marrow Proteoglycan (Prg2) Elisa Kit for detecting Proteoglycan 2, Bone Marrow (Natural Killer Cell Activator, Eosinophil Granule Major Basic Protein) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.9-250ng/ml. Sensitivity: 0.98ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q61878; Prg2. Target Synonyms: Prg2; Mbp-1; Bone marrow proteoglycan; BMPG; Proteoglycan 2) [Cleaved into: Eosinophil granule major basic protein; EMBP; MBP)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8432