Learn More
Biomatik Corporation Mouse Bone morphogenetic protein 6, BMP-6 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36422
Encompass_Preferred
Mouse Bone Morphogenetic Protein 6, Bmp-6 Elisa Kit for detecting Bone Morphogenetic Protein 6 in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P20722; Bmp6. Target Synonyms: Bmp6; Bmp-6; Vgr1Bone morphogenetic protein 6; BMP-6; VG-1-related protein; VGR-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8433