Learn More
Biomatik Corporation Mouse Bone sialoprotein, BSP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36430
Encompass_Preferred
Mouse Bone Sialoprotein , Bsp Elisa Kit for detecting Bone Sialoprotein , Bsp in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P10923; Spp1. Target Synonyms: Spp1; Eta-1; Op; Spp-1; Osteopontin; 2AR; Bone sialoprotein 1; Calcium oxalate crystal growth inhibitor protein; Early T-lymphocyte activation 1 protein; Minopontin; Secreted phosphoprotein 1; SPP-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8434