Learn More
Biomatik Corporation Mouse Bone-specific alkaline phosphatase (BALP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36431
Encompass_Preferred
Mouse Bone-Specific Alkaline Phosphatase (Balp) Elisa Kit for detecting Bone-Specific Alkaline Phosphatase in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.25-80ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P09242; Alpl. Target Synonyms: Alpl; Akp-2; Akp2; Alkaline phosphatase; tissue-nonspecific isozyme; AP-TNAP; TNSALP; EC 3.1.3.1; Alkaline phosphatase 2; Alkaline phosphatase liver/bone/kidney isozyme. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8435