Learn More
Biomatik Corporation Mouse chemokine (C-X-C motif) ligand 17 (CXCL17) ELISA Kit, 96 tests
Catalog No. 501498438
Encompass_Preferred
Mouse Chemokine (C-X-C Motif) Ligand 17 (Cxcl17) Elisa Kit for detecting Chemokine (C-X-C Motif) Ligand 17 (Cxcl17) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 37.5-2400pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q5UW37; Cxcl17. Target Synonyms: Cxcl17; Vcc1C-X-C motif chemokine 17; 6-Cys CXCL17; 13.6 kDa protein; VEGF coregulated chemokine 1) [Cleaved into: 4-Cys CXCL17]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8438 Supplier Biomatik Corporation Supplier No. EKC36537
Mouse Chemokine (C-X-C Motif) Ligand 17 (Cxcl17) Elisa Kit for detecting Chemokine (C-X-C Motif) Ligand 17 (Cxcl17) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 37.5-2400pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q5UW37; Cxcl17. Target Synonyms: Cxcl17; Vcc1C-X-C motif chemokine 17; 6-Cys CXCL17; 13.6 kDa protein; VEGF coregulated chemokine 1) [Cleaved into: 4-Cys CXCL17]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.