Learn More
Biomatik Corporation Mouse Ciliary Neurotrophic Factor, CNTF ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36558
Encompass_Preferred
Mouse Ciliary Neurotrophic Factor, Cntf Elisa Kit for detecting Ciliary Neurotrophic Factor in serum, plasma, tissue homogenates, cell culture supernates. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P51642; Cntf. Target Synonyms: CntfCiliary neurotrophic factor; CNTF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8441