Learn More
Biomatik Corporation Mouse coagulation factor , F ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36571
Encompass_Preferred
Mouse Coagulation Factor V, Fv Elisa Kit for detecting Coagulation Factor V, FV in serum, plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5000ng/ml. Sensitivity: 19.5ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O88783; F5. Target Synonyms: F5Coagulation factor V; Activated protein C cofactor) [Cleaved into: Coagulation factor V heavy chain; Coagulation factor V light chain]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8443