Learn More
Biomatik Corporation Mouse Cold-inducible RNA-binding protein (CIRBP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36584
Encompass_Preferred
Mouse Cold-Inducible Rna-Binding Protein (Cirbp) Elisa Kit for detecting Cold Inducible Rna Binding Protein in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P60824; Cirbp. Target Synonyms: Cirbp; CirpCold-inducible RNA-binding protein; A18 hnRNP; Glycine-rich RNA-binding protein CIRP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8444