Learn More
Biomatik Corporation Mouse Collagen alpha-1 (I) chain (COL1A1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36586
Encompass_Preferred
Mouse Collagen Alpha-1 (I) Chain (Col1A1) Elisa Kit for detecting Collagen, Type I, Alpha 1 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P11087; Col1a1. Target Synonyms: Col1a1; Cola1Collagen alpha-1 (I) chain; Alpha-1 type I collagen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8445