Learn More
Biomatik Corporation Mouse Complement 5, C5 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36627
Encompass_Preferred
Mouse Complement 5, C5 Elisa Kit for detecting Complement 5, C5 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.25-16ng/ml. Sensitivity: 0.06ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P06684; C5. Target Synonyms: C5; HcComplement C5; Hemolytic complement) [Cleaved into: Complement C5 beta chain; Complement C5 alpha chain; C5a anaphylatoxin; Complement C5 alpha' chain]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8449