Learn More
Biomatik Corporation Mouse Complement C1q subcomponent subunit A (C1QA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36628
Encompass_Preferred
Mouse Complement C1Q Subcomponent Subunit A (C1Qa) Elisa Kit for detecting Complement Component 1, Q Subcomponent, A Chain in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ug/ml. Sensitivity: 0.039ug/mL. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P98086; C1qa. Target Synonyms: C1qaComplement C1q subcomponent subunit A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8450